Learn More
Cole-Parmer™ Nickel-plated Brass Luer Fittings
Click to view available options
For Use With:
Female luer lock
Male Luer Lock
Includes:
1/16 in. Hose Barb
10-32 UNF Thread
Unit Size:
Each
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Nickle Plated Brass |
| For Use With | Female luer lock |
| For Use With (Application) | Harsh chemical environments |
| Packaging Quantity | 1 Pack |
| Includes | 1/16 in. Hose Barb |
| Product Type | Luer Fitting |