Learn More
Usbeck Laborgeraete™ Nickel Plated Brass Hoffman Pinchcocks
10.15 € - 19.95 €
Product Code | Brand | Width (Metric) | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Width (Metric) | Preis | Menge & Verfügbarkeit | |||||
10068260
|
Usbeck Laborgeraete™
5513 |
25 mm |
10.70 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10527351
|
Usbeck Laborgeraete™
5514 |
30 mm |
11.35 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10708921
|
Usbeck Laborgeraete™
5515 |
40 mm |
19.95 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10313751
|
Usbeck Laborgeraete™
5511 |
17 mm |
10.15 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10323751
|
Usbeck Laborgeraete™
5512 |
20 mm |
10.45 €
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Spezifikation
Each | |
Hoffman Pinchcock | |
Nickel-plated Brass |