missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, 98%

Product Code. 16675234 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
6.42 kg
Packaging:
Glass bottle
Unit Size:
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
16675234 6.42 kg - -
10414905 50 g Glass bottle 50g
10222463 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16675234 Supplier Thermo Scientific Acros Supplier No. 387320000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nickel(II) chloride, 98%, Cl2Ni, CAS Number-7718-54-9, 250g, 97.5% min. (Argentometry), 100.0, CHEBI:34887, Brown to Orange, 231-743-, 129.6, GHS Signal Word: Danger | CAS: 7718-54-9 | Cl2Ni | 129.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-54-9
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887

Specifications

Melting Point 1001.0°C
Color Brown to Orange
CAS Min % 97.5
CAS Max % 100.0
Physical Form Powder
Quantity 6.42 kg
Assay Percent Range 97.5% min. (Argentometry)
Molecular Formula Cl2Ni
Linear Formula NiCl2
Merck Index 15, 6590
Solubility Information Solubility aq. soln.: slightly soluble
SMILES [Cl-].[Cl-].[Ni++]
IUPAC Name nickel(2+) dichloride
Molecular Weight (g/mol) 129.59
Formula Weight 129.6
Percent Purity 98%
Chemical Name or Material Nickel(II) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.