missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Fe/Ni/Cr, Alloy 825, N08825, ARMI∣MBH
Description
Nickel, Fe/Ni/Cr, Alloy 825, N08825, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Zirconium/Boron/Carbon/Phosphorus/Vanadium/Cobalt/Aluminium/Silicon/Manganese/Titanium/Copper/Molybdenum/Chromium/Iron/Nickel/Magnesium/Niobium |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zirconium 0.0021 %/Boron 0.0028 %/Carbon 0.015 %/Phosphorus 0.0189 %/Vanadium 0.038 %/Cobalt 0.0646 %/Aluminium 0.149 %/Silicon 0.232 %/Manganese 0.499 %/Titanium 1.192 %/Copper 1.87 %/Molybdenum 3.01 %/Chromium 21.94 %/Iron 31.82 %/Nickel 39.12 %/Magnesium (0.003) %/Niobium (0.007) % |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?