missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr, Alloy 500, N07500, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
Each
Description
Nickel, Ni/Cr, Alloy 500, N07500, 100g
Specifications
Specifications
| Chemical Name or Material | Tin/Oxygen/Nitrogen/Phosphorus/Manganese/Magnesium/Vanadium/Zirconium/Boron/Tantalum/Tungsten/Silicon/Niobium/Carbon/Iron/Aluminium/Titanium/Molybdenum/Cobalt/Chromium/Nickel/Copper |
| CAS | 7440-02-0 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Tin 0.0002 %/Oxygen 0.0005 %/Nitrogen 0.0007 %/Phosphorus 0.001 %/Manganese 0.002 %/Magnesium 0.0023 %/Vanadium 0.004 %/Zirconium 0.008 %/Boron 0.009 %/Tantalum 0.01 %/Tungsten 0.013 %/Silicon 0.02 %/Niobium 0.022 %/Carbon 0.079 %/Iron 0.086 %/Aluminium 3.02 %/Titanium 3.02 %/Molybdenum 3.51 %/Cobalt 16.99 %/Chromium 18.47 %/Nickel 54.8 %/Copper (0.001) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction