missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr/Mo, Alloy 725, N07725, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Ni/Cr/Mo, Alloy 725, N07725, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Tin/Boron/Phosphorus/Magnesium/Tantalum/Carbon/Nitrogen/Vanadium/Tungsten/Manganese/Cobalt/Silicon/Copper/Aluminium/Titanium/Niobium/Iron/Molybdenum/Chromium/Nickel/Antimony/Bismuth/Calcium/Lead/Arsenic/Zinc/Zirconium/Rhenium/Yttrium/Hafnium/Oxygen |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.0011 %/Boron 0.0023 %/Phosphorus 0.0047 %/Magnesium 0.005 %/Tantalum 0.006 %/Carbon 0.008 %/Nitrogen 0.008 %/Vanadium 0.013 %/Tungsten 0.04 %/Manganese 0.076 %/Cobalt 0.077 %/Silicon 0.084 %/Copper 0.103 %/Aluminium 0.22 %/Titanium 1.55 %/Niobium 3.47 %/Iron 7.47 %/Molybdenum 7.9 %/Chromium 20.5 %/Nickel 58.4 %/Antimony (<0.0001) %/Bismuth (<0.0001) %/Calcium (<0.0010) %/Lead (<0.0010) %/Arsenic (<0.0020) %/Zinc (<0.0030) %/Zirconium (<0.0050) %/Rhenium (<0.0100) %/Yttrium (<0.0100) %/Hafnium (<0.0102) %/Oxygen (0.0005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction