Learn More
Invitrogen™ Nicarbazin Polyclonal Antibody
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  Invitrogen™ PA175315
Product Code. 11504370
  • 732.00 € / 1mg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Working Concentration: 0.078 μg/mL. Sensitivity: 10 ng/mL Nicarbazin produces 55% inhibition in a competitive ELISA employing Nicarbazin polyclonal antibody.
Nicarbazin is an equimolar complex of 4,4'-Dinitrocarbanilide and 2-Hydroxy-4,6-dimethylpyrimidine. It is used as a coccidiostat for poultry. [National Center for Biotechnology Information. PubChem Compound Database; CID=9507].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nicarbazin
Polyclonal
PBS with 0.09% sodium azide; pH 7.1
Sheep
purified
RUO
Chemical
Antibody
IgG
ELISA
Unconjugated
C19H18N6O6; Nicarbazine; Nicrazin; Nicrazine
Nitrobenzene(4)-BTG.
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications