missing translation for 'onlineSavingsMsg'
Learn More

Nicainoprol, MedChemExpress

Product Code. p-200100884
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485817 5 mg 5mg
16400679 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16485817 Supplier Medchem Express Supplier No. HY100572

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nicainoprol is a fast-sodium-channel blocking drug, which is a potent antiarrhythmic agent.

Nicainoprol is a fast-sodium-channel blocking drug, which is a potent antiarrhythmic agent.

Specifications

Chemical Name or Material Nicainoprol
CAS 76252-06-7
Color White
Purity Grade Notes Research
Molecular Formula C21H27N3O3
Quantity 5 mg
Synonym RU-42924
Solubility Information DMSO : ≥ 29 mg/mL (78.49 mM)
SMILES O=C(N1CCCC2=C1C(OCC(O)CNC(C)C)=CC=C2)C3=CC=CN=C3
Molecular Weight (g/mol) 369.46
Formula Weight 369.46
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.