missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NFATC4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11571432
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11571432 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11571432 Supplier Invitrogen™ Supplier No. PA1021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-021 detects nuclear factor of activated T-cells 3 (NFAT3) from human cells. PA1-021 has been successfully used in Western blot immunohistochemistry, immunofluorescence, and immunoprecipitation procedures. By Western blot, this antibody detects an ~150 kDa protein representing NFAT3 in HEK293 cells transfected with the human NFAT3 gene. Immunohistochemical staining of NFAT3 in HEK293 cells transfected with the human NFAT3 gene results in cytoplasmic staining. The PA1-021 immunogen is a synthetic peptide corresponding to residues C R(887) D L S G F P A P P G E E P P A(902) of human NFAT3. This sequence is 94% conserved with mouse NFAT3. The PA1-021 immunizing peptide (Cat. # PEP-122) is available for use in neutralization and control experiments.

This protein is an adapter or scaffold protein, which participates in the assembly of numerous protein complexes and is involved in several cellular processes such as cell fate determination, cytoskeletal organization, repression of gene transcription, mitosis, cell-cell adhesion, cell differentiation, proliferation and migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC4
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Nfatc4
Gene Accession No. Q14934
Gene Alias 3110041H08Rik; AW107667; AW546455; Nfat3; NF-AT3; Nfatc4; NF-ATc4; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 4; nuclear factor of activated T-cells 4; nuclear factor of activated T-cells, cytoplasmic 4; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4; T-cell transcription factor NFAT3; transcription complex subunit NF-ATc4
Gene Symbols Nfatc4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C R(887) D L S G F P A P P G E E P P A(902) of human NFAT3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4776
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.