missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Nexinhib20

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
Nexinhib20 is a Rab27 inhibitor (IC50 = 330 nM); inhibits Rab27a-JFC1 binding. Selectively inhibits neutrophil exocytosis, with no significant effect on other important innate immune responses including phagocytosis. Suppresses extracellular production of superoxide anion and cytochrome b plasma membrane expression. Also inhibits upregulation of adhesion molecules CD11b and CD66b. Displays in vivo anti-inflammatory activity.
Specifications
Specifications
| Chemical Name or Material | 4,4-Dimethyl-1-(3-nitrophenyl)-2-(1H-1,2,4-triazol-1-yl)-1-penten-3-one |
| CAS | 331949-35-0 |
| Quantity | 5 mg |
| Molecular Formula | C15H16N4O3 |
| Formula Weight | 300.31 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction