missing translation for 'onlineSavingsMsg'
Learn More

Nevirapine, MedChemExpress

Product Code. 16450407
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16450407

Brand: Medchem Express HY10570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nevirapine is a non-nucleoside inhibitor of HIV-1 reverse transcriptase used to treat and prevent HIV/AIDS; with a Ki of 270 μM.

Nevirapine is a non-nucleoside inhibitor of HIV-1 reverse transcriptase used to treat and prevent HIV/AIDS

Specifications

Chemical Name or Material Nevirapine
CAS 129618-40-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H14N4O
Quantity 50 mg
Synonym BI-RG 587 NSC 641530 NVP
Solubility Information DMSO : 14.29 mg/mL (53.66 mM; Need ultrasonic)
SMILES O=C1C2=C(N=CC=C2)N(C3CC3)C4=NC=CC(C)=C4N1
Molecular Weight (g/mol) 266.3
Formula Weight 266.3
Percent Purity 99.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.