missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropilin-1 (CD304) Mouse anti-Human, BV421, Clone: U21-1283, BD Horizon™
Mouse Monoclonal Antibody
178.69 € - 373.00 €
Specifications
| Antigen | Neuropilin-1 (CD304) |
|---|---|
| Clone | U21-1283 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15844629
|
BD Biosciences
565954 |
25 Tests |
179.00 € 178.69 € / Each Save 0.31 € 0.17% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15834629
|
BD Biosciences
565953 |
100 Tests |
373.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.Specifications
| Neuropilin-1 (CD304) | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Recombinant Human Neuropilin-1 | |
| Primary | |
| Vol. Per test: 5μL |
| U21-1283 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Neuropilin; NRP; NP1; Blood dendritic cell antigen 4; BDCA-4; VEGF165R | |
| IgG2b κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title