Learn More
Neuronatin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26905-1-AP-150UL
 View more versions of this product
Product Code. 16875235
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
NNAT may participate in the maintece of segment identity in the hindbrain and pituitary development, and maturation or maintece of the overall structure of the nervous system. may function as a regulatory subunit of ion channels.
Specifications
Specifications
Neuronatin
Polyclonal
Unconjugated
NNAT
neuronatin, NNAT, Peg5
Rabbit
Antigen Affinity Chromatography
RUO
4826
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q16517
NNAT
Neuronatin Fusion Protein Ag25408
150 μL