Learn More
Neuronatin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26905-1-AP-150UL
This item is not returnable.
View return policy
Description
NNAT may participate in the maintece of segment identity in the hindbrain and pituitary development, and maturation or maintece of the overall structure of the nervous system. may function as a regulatory subunit of ion channels.Specifications
| Neuronatin | |
| Polyclonal | |
| Unconjugated | |
| NNAT | |
| neuronatin, NNAT, Peg5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4826 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16517 | |
| NNAT | |
| Neuronatin Fusion Protein Ag25408 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |