Description
Neuronal Pentraxin R/NPTXR Polyclonal specifically detects Neuronal Pentraxin R/NPTXR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Neuronal Pentraxin R/NPTXR |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Gene Symbols | NPTXR |
| Host Species | Rabbit |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mL |
| Isotype | IgG |