Learn More
NEURL Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99299PCP
Product Code. 18627775
  • 407.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
NEURL Polyclonal antibody specifically detects NEURL in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence