Learn More
Neuregulin-1 beta 1/NRG1 beta 1 EGF Domain Mouse anti-Human, Clone: 147705, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB3771-500
Description
Neuregulin-1 beta 1/NRG1 beta 1 Monoclonal specifically detects Neuregulin-1 beta 1/NRG1 beta 1 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Neuregulin-1 beta 1/NRG1 beta 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, neuregulin 1, Neuregulin1, Neuregulin1 beta 1, NRG1, NRG1-IT2, SMDF | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3084 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 147705 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| AAA58639 | |
| NRG1 | |
| E. coli-derived recombinant human NRG1-beta 1/HRG1-beta 1 Ser2-Lys246 Accession # AAA58639 | |
| 500 μg | |
| Primary | |
| Detects human NRG1-beta 1 in ELISAs. Recognizes an epitope found in the EGF-like domain of NRG1-beta 1 (amino acids 179-246). It also detects recombinant human SMDF (Sensory and Motor neuron-Derived Factor), but it does not recognize the EGF-like domain of human NRG1-alpha (amino acids 177-241). The epitope is present within amino acids 213-230 of human NRG1-beta 1. This segment is present in all NRG1 beta isoforms, GGF-2 (glial growth factor), and SMDF, but it is absent from NRG1-alpha , NDF43, and NRG1-gamma . | |
| Human | |
| Purified |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.