missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NETO1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00 € - 507.00 €
Specifications
| Antigen | NETO1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696015
|
Novus Biologicals
NBP2-41134-0.025mg |
0.025 mg |
178.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18124829
|
Novus Biologicals
NBP2-41134 |
0.1 mg |
507.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NETO1 Polyclonal specifically detects NETO1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NETO1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1, BTCL1BCTL1neuropilin and tolloid-like protein 1, FLJ41325, neuropilin (NRP) and tolloid (TLL)-like 1 | |
| NETO1 | |
| IgG | |
| Affinity Purified | |
| 60 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| PBS with 0.02% Sodium Azide | |
| 81832 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human NETO1. The immunogen is located within amino acids 350 - 400 of NETO1. Amino Acid Squence: KQPRKKYVQRKSDFDQTV | |
| Primary | |
| NETO1 antibody is predicted to not cross-react with other NETO protein family members. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title