missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes

Product Code. 30245276
Click to view available options
No. of Reactions:
384 x 2 Reactions
96 x 2 Reactions
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30245276

Brand: Invitrogen™ N1003848

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Neon NxT Electroporation System 100-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station.

The Neon NxT Electroporation System 100-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. No. MPK5000) with 8-channel pipette and pipette station (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Use this kit for transfection volumes of 100 μL, containing 5 x 105 to 2 x 106 adherent cells or 1 x 106 to 5 x 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be plated into a multi-well culture dish.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Box 1 includes:
• Resuspension R Buffer, 2 x 80 mL
• Resuspension T Buffer, 2 x 80 mL
• Resuspension Genome Editing Buffer, 2 x 80 mL
• Electrolytic E100 Buffer, 2 x 100 mL

Store at 4°C.

Box 2 includes:
• Reaction tips, 384 tips
• Electroporation 8-channel tubes, 16 tubes

Store at room temperature.
Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Electroporation Pipette Tip
Transfection Technique Electroporation
For Use With (Equipment) Neon™ NxT Electroporation System
For Use With (Application) Transfection
No. of Reactions 384 x 2 Reactions
Quantity 384 x 2 Reactions
Volume (Metric) Sample 100 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.