Learn More
Melford Neomycin Sulfate
Description
Neomycin Sulfate is an antibiotic that is derived from the bacterium Streptomyces fradiae and belongs to the aminoglycoside class of antibiotics.
In molecular biology, Neomycin Sulfate is commonly used as a selective agent in genetic engineering experiments and cell culture applications. It is often included in growth media or buffers to prevent the growth of bacteria and ensure the purity of cell lines.
Neomycin Sulfate functions by inhibiting protein synthesis in bacteria, leading to cell death. Neomycin Sulfate is also employed in microbiology research. It can be used to selectively isolate and identify bacteria or as a control for susceptibility testing in antimicrobial resistance studies.
Specifications
Specifications
| Chemical Name or Material | Neomycin Sulfate |
| CAS | 1405-10-3 |
| Quantity | 25 g |
| Molecular Formula | C23H46N6O13-3H2OSO4 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.