missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neogenin Antibody (RM0124-3G55), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22501CL1
This item is not returnable.
View return policy
Description
Neogenin Monoclonal antibody specifically detects Neogenin in Mouse samples. It is validated for Western Blot
Specifications
| Neogenin | |
| Monoclonal | |
| CoraFluor 1 | |
| HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGNDKFZp547A066 | |
| Mouse recombinant protein of Neogenin | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0124-3G55 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 4756 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction