missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEK6 Recombinant Rabbit Monoclonal Antibody (1V1X8)
GREENER_CHOICE

Product Code. 17236953 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17236953 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17236953 Supplier Invitrogen™ Supplier No. MA538077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Samples: 293T, K-562, Mouse lung, Mouse brain, Rat brain Immunogen sequence: ETTAAHSLVG TPYYMSPERI HENGYNFKSD IWSLGCLLYE MAALQSPFYG DKMNLFSLCQ KIEQCDYPPL PGEHYSEKLR ELVSMCICPD PHQRPDIGYV H.

NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition. The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1V1X8
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NEK6
Gene Accession No. P59895, Q9ES70, Q9HC98
Gene Alias 1300007C09Rik; Nek6; Never in mitosis A-related kinase 6; NIMA (never in mitosis gene a)-related expressed kinase 6; NIMA (never in mitosis gene a)-related kinase 6; NIMA related kinase 6; NIMA-related expressed kinase 6; NIMA-related kinase 6; nimA-related protein kinase 6; Protein kinase SID6-1512; putative serine-threonine protein kinase; RP11-101K10.6; serine/threonine-protein kinase Nek6; SID; SID6-1512
Gene Symbols NEK6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-300 of human NEK6 (Q9HC98).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10783, 360161, 59126
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.