Learn More
Aalborg™ Needle Valve for Brass Flowmeter
Product Code. 11812400
Click to view available options
Airflow Rate:
5000 mL/min.
60000 mL/min.
Waterflow Rate:
3500mL/min.
350mL/min.
Unit Size:
Each
Product Code. 11812400
Brand: Aalborg™ VCHBB
Only null left
Add to basket
Add to basket
These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes. Aalborg™ Needle Valve for Brass Flowmeter is designed for controlling a broad range of flow rates of liquids and gases.
  • Available in three conveniently overlapping orifice-needle sizes.
  • These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes.
  • Valves are offered in straight (T) and 90 degree (L) flow patterns. All valves are supplied with 1/8in. FNPT inlet and outlet ports.
  • Valve cartridges are also interchangeable with built-in valves of Aalborg’s series of P, T, S, and G flow meter product line.
  • Bubble tight shutoff
Specifications
For Use With (Equipment) 150 mm Gas/Liquid Flowmeter
Type Standard Valve
Airflow Rate 60000 mL/min.
Waterflow Rate 3500mL/min.
Max. Operating Temperature (Metric) 82°C