missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NEDD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30524
This item is not returnable.
View return policy
Description
NEDD4 Polyclonal antibody specifically detects NEDD4 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| NEDD4 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1 | |
| between 825 and 875 | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Endocrinology, Immunology, Neuroscience, Signal Transduction | |
| 4734 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction