missing translation for 'onlineSavingsMsg'
Learn More

NE 10790, MedChemExpress

Product Code. 16487036
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487036

Brand: Medchem Express HY16011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NE 10790, a poor farnesyl pyrophosphate synthase inhibitor, is a phosphonocarboxylate analogue of the potent bisphosphonate risedronate and is a weak antiresorptive agent.

NE 10790, a poor farnesyl pyrophosphate synthase inhibitor, is a phosphonocarboxylate analogue of the potent bisphosphonate risedronate and is a weak antiresorptive agent.

Specifications

Chemical Name or Material NE 10790
Physical Form Solid
CAS 152831-36-2
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H10NO6P
Synonym 3-PEHPC
Solubility Information 0.1 M NaOH : 20 mg/mL (80.93 mM; ultrasonic and adjust pH to 6 with NaOH) ∣H2O : 5 mg/mL (20.23 mM; ultrasonic and adjust pH to 6 with NaOH)
SMILES OC(C(O)=O)(P(O)(O)=O)CC1=CC=CN=C1
Molecular Weight (g/mol) 247.14
Formula Weight 247.14
Percent Purity 98.28%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.