missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: LLKDEIVDHW RGPEGHPLHE VLLEQAKVVW QFRGKEDLDK WTVTSDKTIG GRSEVFLKMG KNNQSALLYG TLSSEAPQDG ESTRSGYCAM ISRIPRGAF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.
The NDUFAF1 gene encodes the human homolog of a Neurospora crassa Cia30 protein involved in the assembly of complex I.
Specifications
Specifications
| Antigen | NDUFAF1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NDUFAF1 |
| Gene Accession No. | Q9Y375 |
| Gene Alias | 2410001M24Rik; CGI65; CGI-65; CIA30; complex I intermediate-associated protein 30, mitochondrial; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1; NADH dehydrogenase (ubiquinone) 1 assembly factor 1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 1; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1; NADH:ubiquinone oxidoreductase complex assembly factor 1; NADH-ubiquinone oxidoreductase 1 alpha subcomplex, assembly factor 1; Ndufaf1 |
| Gene Symbols | NDUFAF1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
