Learn More
NDUFA4L2 Mouse anti-Human, Clone: OTI5C10, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810602S
Description
NDUFA4L2 Monoclonal antibody specifically detects NDUFA4L2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NDUFA4L2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NRX3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 56901 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C10 | |
| Unconjugated | |
| NDUFA4L2 | |
| BC064011; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4-like 2; NADH:ubiquinone oxidoreductase MLRQ subunit homolog; NADH-ubiquinone oxidoreductase MLRQ subunit homolog; NDUFA4, mitochondrial complex associated like 2; NDUFA4L2; NUOMS | |
| Full length human recombinant protein of human NDUFA4L2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.