missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDRG1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB300-839
This item is not returnable.
View return policy
Description
NDRG1 Polyclonal specifically detects NDRG1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| NDRG1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CAP43, CMT4D, Differentiation-related gene 1 protein, DRG-1, DRG1HMSNL, NDR1, Nickel-specific induction protein Cap43, NMSL, N-myc downstream regulated 1, N-myc downstream-regulated gene 1 protein, protein NDRG1, protein regulated by oxygen-1, PROXY1, Reducing agents and tunicamycin-responsive protein, RIT42, RTPGC4, TARG1, TDD5, tunicamycin-responsive protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The variants (NP_006087.2; NP_001128714.1) encode the same protein | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 μg/mL, Immunohistochemistry 0.5 - 2 μg/mL, Immunohistochemistry-Paraffin 0.5-2 μg/mL, Peptide ELISA detection limit 1:32000 | |
| NP_006087. | |
| NDRG1 | |
| Peptide with sequence GNSAGPKSMEVSC corresponding to C-Terminus according to NP_006087.2, NP_001128714.1. | |
| 0.1 mg | |
| Cancer | |
| 10397 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction