missing translation for 'onlineSavingsMsg'
Learn More

NCT-501, MedChemExpress

Product Code. 16417628
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16417628

Brand: Medchem Express HY18768

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NCT-501 is a potent and selective theophylline-based inhibitor of aldehyde dehydrogenase 1A1 (ALDH1A1), inhibits hALDH1A1 with IC50 of 40 nM, typically shows better selectivity over other ALDH isozymes and other dehydrogenases (hALDH1B1, hALDH3A1, and hALDH2, IC50 >57 μM).

NCT-501 is a potent and selective theophylline-based inhibitor of aldehyde dehydrogenase 1A1 (ALDH1A1), inhibits hALDH1A1 with IC50 of 40 nM, typically shows better selectivity over other ALDH isozymes and other dehydrogenases (hALDH1B1, hALDH3A1, and hALDH2, IC50 >57 μM).

Specifications

Chemical Name or Material NCT-501
CAS 1802088-50-1
Color White
Purity Grade Notes Research
Molecular Formula C21H32N6O3
Quantity 10 mg
Solubility Information DMSO : ≥ 28 mg/mL (67.22 mM)
SMILES O=C(N1C)N(C)C2=C(N(CCC(C)C)C(CN3CCN(C(C4CC4)=O)CC3)=N2)C1=O
Molecular Weight (g/mol) 416.52
Formula Weight 416.52
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.