Learn More
NCOA2 Monoclonal Antibody (OTI7G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805128S
Description
NCOA2 Monoclonal antibody specifically detects NCOA2 in Human samples. It is validated for Western BlotSpecifications
| NCOA2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15596 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10499 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7G5 | |
| Unconjugated | |
| NCOA2 | |
| bHLHe75; c-fgr; Class E basic helix-loop-helix protein 75; c-src2; D1Ertd433e; FLJ43153; Glucocorticoid receptor-interacting protein 1; glucocorticoid receptor-interacting protein-1; Grip1; GRIP-1; hTIF2; I79_019839; KAT13C; MGC75096; NCOA2; NCoA-2; Nuclear receptor coactivator 2; p160 steroid receptor coactivator 2; p55c-fgr; p58c-fgr; SRC2; SRC-2; Steroid receptor coactivator 2; TIF 2; Tif2; transcriptional intermediary factor 2 | |
| Human recombinant protein fragment corresponding to amino acids 1-280 of human NCOA2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.