missing translation for 'onlineSavingsMsg'
Learn More

NCGC00244536, MedChemExpress

Product Code. 16479986
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16479986

Brand: Medchem Express HY101799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NCGC00244536 is a potent KDM4B inhibitor with an IC50 of 10 nM.

NCGC00244536 is a potent KDM4B inhibitor with an IC50 of 10 nM.

Specifications

Chemical Name or Material NCGC00244536
CAS 2003260-55-5
Color Yellowish Green
Purity Grade Notes Research
Molecular Formula C25H22N2O2
Quantity 50 mg
Synonym KDM4B Inhibitor B3
Solubility Information DMSO : 36.67 mg/mL (95.88 mM; Need ultrasonic)
SMILES O=C(NCCCC1=CC=CC=C1)C2=CC=CC(C3=CC(O)=C4N=CC=CC4=C3)=C2
Molecular Weight (g/mol) 382.45
Formula Weight 382.45
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.