missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCDN Polyclonal Antibody
GREENER_CHOICE

Product Code. 16550822
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16550822 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16550822 Supplier Invitrogen™ Supplier No. PA597996

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NCDN Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Probably involved in signal transduction, in the nervous system, via increasing cell surface localization of GRM5 and positively regulating its signaling. Required for the spatial learning process. Acts as a negative regulator of Ca(2+)-calmodulin-dependent protein kinase 2 (CaMK2) phosphorylation. May play a role in modulating melanin-concentrating hormone-mediated functions via its interaction with MCHR1 that interferes with G protein-coupled signal transduction. May be involved in bone metabolism. May also be involved in neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCDN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NCDN
Gene Accession No. O35095, Q9UBB6
Gene Alias AU042419; Kiaa0607; minisatellite 10ae detected by probe MMS10; mKIAA0607; MMS10-AE; Ms10ae; M-Sema F-associating protein of 75 kDa; NCDN; Neurite outgrowth-related protein from the rat brain; neurochondrin; neurochondrin-1; neurochondrin-2; Norbin; Sfap75; similar to neurochondrin
Gene Symbols NCDN
Host Species Rabbit
Immunogen Recombinant Human Neurochondrin protein (403-516AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23154, 89791
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.