missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCCRP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571715
This item is not returnable.
View return policy
Description
NCCRP1 Polyclonal Antibody for Western Blot
Promotes cell proliferation.
Specifications
| NCCRP1 | |
| Polyclonal | |
| Unconjugated | |
| NCCRP1 | |
| 1190020J12Rik; F-box only protein 50; FBXO50; Gm163; NCC receptor protein 1; NCC receptor protein 1 homolog; NCCRP1; NCCRP-1; nonspecific cytotoxic cell receptor protein 1 homolog; Non-specific cytotoxic cell receptor protein 1 homolog; non-specific cytotoxic cell receptor protein 1 homolog (zebrafish) | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 233038, 342897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| G3X9C2, Q6ZVX7 | |
| NCCRP1 | |
| KLH conjugated synthetic peptide between 246-275 amino acids from the C-terminal region of human NCCRP1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction