missing translation for 'onlineSavingsMsg'
Learn More

NCC007, MedChemExpress

Product Code. 16425418
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425418

Brand: Medchem Express HY128677

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NCC007 is a dual casein kinase Iα (CKIα) and δ (CKIδ) inhibitor with IC50s of 1.8 and 3.6 μM, respectively. NCC007 can be used in research of modulating mammalian circadian rhythms.

NCC007 is a dual casein kinase Iα (CKIα) and δ (CKIδ) inhibitor with IC50s of 1.8 and 3.6 μM, respectively. NCC007 can be used in research of modulating mammalian circadian rhythms.

Specifications

Chemical Name or Material NCC007
CAS 2342583-66-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H28F3N7
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (279.33 mM; Need ultrasonic)
SMILES CN(C)CCNC1=NC2=C(N=CN2C(C)C3CC3)C(NCC4=CC(C(F)(F)F)=CC=C4)=N1
Molecular Weight (g/mol) 447.5
Formula Weight 447.5
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.