Learn More
NCC007, MedChemExpress
Product Code. 16491597
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16491597
Brand: Medchem Express HY128677
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress NCC007 is a dual casein kinase Iα (CKIα) and δ (CKIδ) inhibitor with IC50s of 1.8 and 3.6 μM, respectively. NCC007 can be used in research of modulating mammalian circadian rhythms.
NCC007 is a dual casein kinase Iα (CKIα) and δ (CKIδ) inhibitor with IC50s of 1.8 and 3.6 μM, respectively. NCC007 can be used in research of modulating mammalian circadian rhythms.
Specifications
Chemical Name or Material NCC007
CAS 2342583-66-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H28F3N7
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (279.33 mM; Need ultrasonic)
SMILES CN(C)CCNC1=NC2=C(N=CN2C(C)C3CC3)C(NCC4=CC(C(F)(F)F)=CC=C4)=N1
Molecular Weight (g/mol) 447.5
Formula Weight 447.5
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation