Learn More
NBN Mouse anti-Human, Unconjugated, Clone: 1E11E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66980-1-IG-20UL
Description
The NBN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NBN. This antibody recognizes human antigen. The NBN antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| NBN | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunofluorescence, ELISA | |
| 1E11E10 | |
| Unconjugated | |
| 20 μL | |
| 4683 | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.