Learn More
NBI-74330, MedChemExpress
Product Code. 16489037
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16489037
Brand: Medchem Express HY15320
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress NBI-74330 is a potent antagonist for CXCR3, and exhibits potent inhibition of (125I)CXCL10 and (125I)CXCL11 specific binding with Ki of 1.5 and 3.2 nM, respectively.
NBI-74330 is a potent antagonist for CXCR3, and exhibits potent inhibition of (125I)CXCL10 and (125I)CXCL11 specific binding with Ki of 1.5 and 3.2 nM, respectively.
Specifications
Chemical Name or Material NBI-74330
CAS 855527-92-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H27F4N5O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (165.13 mM; Need ultrasonic)
SMILES O=C(N([C@@H](C1=NC2=NC=CC=C2C(N1C3=CC=C(OCC)C=C3)=O)C)CC4=CC=CN=C4)CC5=CC=C(F)C(C(F)(F)F)=C5
Molecular Weight (g/mol) 605.58
Formula Weight 605.58
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation