missing translation for 'onlineSavingsMsg'
Learn More

NBD-556, MedChemExpress

Product Code. 16418218
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16418218

Brand: Medchem Express HY76648

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NBD-556, a CD4 mimetic, is a potent HIV-1 entry inhibitor that blocks the gp120-CD4 interaction. NBD-556 shows potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.

NBD-556, a CD4 mimetic, is a potent HIV-1 entry inhibitor that blocks the gp120-CD4 interaction. NBD-556 shows potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.

Specifications

Chemical Name or Material NBD-556
CAS 333353-44-9
Color White
Purity Grade Notes Research
Molecular Formula C17H24ClN3O2
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (98.66 mM; Need ultrasonic)
SMILES O=C(C(NC1CC(C)(NC(C)(C1)C)C)=O)NC2=CC=C(C=C2)Cl
Molecular Weight (g/mol) 337.84
Formula Weight 337.84
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.