missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nav1.7 Antibody (N68/6), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12904AF647
This item is not returnable.
View return policy
Description
Nav1.7 Monoclonal specifically detects Nav1.7 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Nav1.7 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| ETHA, FEB3B, NE-NA, NENAVoltage-gated sodium channel subunit alpha Nav1.7, Neuroendocrine sodium channel, Peripheral sodium channel 1, PN1hNE-Na, sodium channel protein type 9 subunit alpha, Sodium channel protein type IX subunit alpha, sodium channel, voltage-gated, type IX, alpha subunit, voltage-gated sodium channel alpha subunit Nav1.7, voltage-gated, type IX, alpha polypeptide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6335 | |
| Human, Mouse, Rat, Hamster | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| S68-6 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| SCN9A | |
| Fusion protein amino acids 1751-1946 (C-terminus) of human Nav1.7 | |
| 0.1 mL | |
| Primary | |
| Detects approx 230kDa. No cross-reactivity against other Nav channels. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction