Learn More
BRAND™ Natural Rubber Stoppers
Shop All Brand Products
Click to view available options
Diameter (Metric) Top:
12 mm
14 mm
22 mm
55 mm
Unit Size:
Each
Pack of 25
Description
- Natural rubber construction
- Red color
- Supplied in packs of 25
Specifications
Specifications
| Height (Metric) | 40 mm |
| Quantity | 25/PK |
| Color | Red |
| Diameter (Metric) Bottom | 47 mm |
| Diameter (Metric) Top | 55 mm |
| Material | Natural Rubber |