missing translation for 'onlineSavingsMsg'
Learn More

Natural Killer Cell Antibody (H25), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18731552 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18731552 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18731552 Supplier Novus Biologicals Supplier No. NBP250526MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Natural Killer Cell Monoclonal antibody specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Radioimmune Assays
TRUSTED_SUSTAINABILITY

Specifications

Antigen Natural Killer Cell
Applications Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone H25
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry, Immunoprecipitation, Radioimmunoassay
Formulation 50mM Sodium Borate
Gene Alias NK Cell
Host Species Mouse
Immunogen HSE-2 cell line
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.