Learn More
Thermo Scientific™ National Unassembled Certified™ Vial Kits
Product Code. 10514315
Click to view available options
Volume (Metric):
1 mL
4 mL
Closure Material:
Bonded PTFE, Silicone
Polyethylene
Closure Type:
Screw Cap
SepCap
Color:
Amber
Unit Size:
Pack of 100
Pack of 200
Product Code. 10514315
Brand: Thermo Scientific™ CERT401599
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
  • Fully lot tested for HPLC and GC analysis
  • Certificate of Conformance in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling
TRUSTED_SUSTAINABILITY
Specifications
Description Certified 1mL Amber Glass
Color Amber
Volume (Metric) 1 mL
Closure Material Polyethylene
Closure Type SepCap