Learn More
Thermo Scientific™ National Storerooms with Target™ Crimp-Top Vials
Click to view available options
Marking Spot:
No
Yes
Unit Size:
Each
Description
Thermo Scientific™ National™ Storerooms with Target Crimp-Top Vials feature your choice of mini (six-drawer) or full size (nine-drawer) storage units complete with vials and caps.
- Mini Storeroom has 6 drawers; each drawer contains 100 packs of vials or septa
- Full size storeroom has 9 deep drawers with a capacity of 2000 vials and seals
- Stackable cabinet constructed of heavy-gauge steel
- With metal shelves and injection-molded drawers
- Storeroom can be restocked as product is consumed
Includes:Prestocked with 3000 each of: clear vials and preassembled clear crimp seals with FEP/red rubber septa
Specifications
Specifications
| Closure Color | Clear |
| Description | NSC Storeroom, Clear I-D Vial |
| Color | Clear |
| Material | PTFE |
| Septum | PTFE/RR Seals |
| Closure Type | Crimp Cap |
| Marking Spot | Yes |