Learn More
Thermo Scientific™ National Scientific Polyethylene SepCap™ Cap
Click to view available options
Type:
12 x 32 Replacement Closure
Closure for Shell Vials
Unit Size:
Pack of 500
Description
- Natural color
- Polyethylene with 0.010 in. built-in septa
Specifications
Specifications
| Color | Natural |
| Material | Polypropylene |
| Type | Closure for Shell Vials |