Learn More
Thermo Scientific™ National Polypropylene and Glastic™ Vials with 8-425 Finish
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
Brand: Thermo Scientific™ C4013-19
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
- Precision formed tips are designed to aid sample recovery for limited-volume applications
- Glastic vial features a glass insert pre-inserted inside a clear TPX vial
Specifications
600µL MicroVials | |
Polypropylene | |
<4 μL | |
8-425 |
Clear | |
600 μL | |
400 μL |