Learn More
Thermo Scientific™ National Economy Vials
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Color:
Amber
Clear
Type:
Clear
Crimp Top Vial
Screw Thread Vial
Snap cap
Volume (Metric) Working:
2 mL
missing translation for 'unitSize'
Pack of 1000
Description
Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.
Specifications
Specifications
| Description | 60180-702 |
| Color | Amber |
| Volume (Metric) | 2 mL |
| Volume (Metric) Working | 2 mL |
| Type | Crimp Top Vial |
| For Use With (Equipment) | Thermo Scientific, Agilent, PE, Shimadzu, Varian |