Learn More
Kautex™ Narrow Neck LDPE Bottles, Natural
Product Code. 15503503
Click to view available options
Capacity (Metric):
3 mL
10 mL
20 mL
50 mL
100 mL
200 mL
250 mL
1000 mL
Unit Size:
Each
Pack of 1500
Pack of 1800
Pack of 3600
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15503503
Brand: Kautex™ 2000070506
Only null left
Add to basket
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Use Kautex™ Narrow Neck LDPE Bottles, Natural in the laboratory as an essential tool with which to conduct research and experiments. Secure them tightly with closures available for purchase separately.
  • Without closure
  • Narrow mouth
  • LDPE construction
  • Metric capacity
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Material LDPE
No. per Pack 1 Ea.
Packaging Bottle
Quantity 1 Ea.
Mouth Narrow
Height (Metric) 140 mm
Product Type Narrow Mouth Bottle
Diameter (Metric) 61.3 mm