Learn More
BRAND™ Narrow Neck Glass Erlenmeyer Flasks
Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
250 mL
300 mL
500 mL
1000 mL
3000 mL
Unit Size:
Each
Pack of 10
Description
- With beaded rim and graduation
- Narrow neck for controlled pouring and dispensing
- Differing package counts according to capacity
Specifications
Specifications
| Certifications/Compliance | DIN ISO 1773 |
| Diameter (Metric) Outer Neck | 34 mm |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass (3.3) |
| Height (Metric) | 180 mm |
| Neck Style | Narrow |
| No. per Pack | 10 |
| Product Type | Erlenmeyer flask |
| Graduated | Yes |
| Diameter (Metric) Outer | 105 mm |