missing translation for 'onlineSavingsMsg'
Learn More

Naringin, MedChemExpress

Product Code. p-200098145
Encompass_Preferred
Click to view available options
Quantity:
200 mg
10 g
Unit Size:
10g
200mg
This item is not returnable. View return policy

Product Code. 16488878

Brand: Medchem Express HYN0153

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Naringin is a major flavanone glycoside obtained from tomatoes, grapefruits, and many other citrus fruits. Naringin exhibits biological properties such as antioxidant, anti-inflammatory, and antiapoptotic activities.

Naringin is a major flavanone glycoside obtained from tomatoes, grapefruits, and many other citrus fruits. Naringin exhibits biological properties such as antioxidant, anti-inflammatory, and antiapoptotic activities.

Specifications

Chemical Name or Material Naringin
CAS 10236-47-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H32O14
Quantity 10 g
Synonym Naringoside
Solubility Information DMSO : 250 mg/mL (430.64 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.72 mM; ultrasonic and warming and heat to 80°C) ∣Ethanol : 1 mg/mL (1.72 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C[C@@H](C2=CC=C(O)C=C2)OC3=CC(O[C@H]4[C@@H]([C@H]([C@@H]([C@@H](CO)O4)O)O)O[C@@]5([H])[C@@H]([C@@H]([C@H]([C@H](C)O5)O)O)O)=CC(O)=C13
Molecular Weight (g/mol) 580.53
Formula Weight 580.53
Percent Purity 98.41%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.