Product Code. 10664922
Shop All Fisher Chemical Products
Click to view available options
Packaging:
Metal can
Plastic Coated Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
10664922
25 L
Metal can
25 L
10558060
2.5 L
Plastic Coated Glass bottle
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
10664922
Supplier
Fisher Chemical
Supplier No.
N/0050/25
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Chemical Identifiers
CAS
8030-30-6
MDL Number
81849
Synonym
benzol, cyclohexatriene, benzole, pyrobenzole, benzine, phenyl hydride, pyrobenzol, benzen, phene, mineral naphtha
Specifications
Color
Colorless
Boiling Point
158°C
Packaging
Metal can
Quantity
25 L
Physical Form
Liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?