Learn More
Macherey-Nagel™ Nanaocolor™ Tube Test
Perform easy, fast measurements. Macherey-Nagel™ Nanocolor™ Tube Tests are bar coded for automatic recognition by Macherey-Nagel photometers. Each test includes exactly pre-dosed reagents in 16mm cuvettes and additional reagents for easy handling, high user safety and maximum precision.
Brand: Macherey-Nagel™ 985088
Additional Details : Weight : 0.10000kg ??product.attributeSet.Transport?? : UN number : 3316 Chem class : 9 Pack group : II
Description
Nanocolor™ tube tests for photometric analysis feature easy handling, making them the first choice for routine, laboratory and process analysis. They are ideally packed in stable boxes with color coded labels, giving all relevant information about the test at one glance. A variety of test kits are available with different measurement ranges for all typical parameters relevant in water and waste water analysis.
- Easy
- Colored pictograms as step-by-step instruction
- Big cuvettes for easy pipetting
- Barcoded cuvettes for automatic test selection
- Safe
- Convenient withdrawal of tubes from the box
- No contact with chemicals
- Reactions based on internationally accepted standard methods
- Reliable
- Precisely pre-dosed reagents
- Adequate test for every application
- Constant high quality from batch to batch
Specifications
Total Nitrogen (N) | |
Orange label | |
Photometric determination | |
Box A: 20 decomposition tubes A, 4g decomposition reagent, 1 tube NANOFIX compensation reagent, 1 orange measuring spoon 85mm Box B: 20 test tubes total Nitrogen TNb 220, 1 test tube with 11mL NO3 / N R2 |
|
20 Pack | |
500 μL | |
30 min. (decomposition), 10 min. (reaction) |
DIN, EN, ISO, EPA and APHA standards | |
Pre-dosed Cuvettes | |
Compatible Macherey-Nagel photometers | |
20 | |
5 - 220mg/L N | |
2,6-Dimethylphenol | |
Cuvette Test |
Safety and Handling
- NANOCOLOR total Nitrogen TNb 220
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : sodium carbinate, potassium peroxydisulphate, 2,6-dimethyl phenol, 2-propanol, o-phosphoric acid, su